Assay Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019936.3 | NP_064320.1 | 2 - 3 | 179 | 116 | |
| GenBank mRNA | AF151637.1 | - | 2 - 3 | 144 | 116 | |
| AK004828.1 | - | 2 - 3 | 172 | 116 | ||
| AK008523.1 | - | 2 - 3 | 164 | 116 | ||
| AK029409.1 | - | 2 - 3 | 167 | 116 | ||
| BC023013.1 | - | 2 - 3 | 153 | 5369938 | 116 | |
| BY123761.1 | - | 2 - 3 | 180 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF151637.1, AK004828.1, AK008523.1, AK029409.1, BC023013.1, NM_019936.3 transcripts.
Set Membership: |
|

Cript
Genomic Map