Assay Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010137.3 | NP_034267.3 | 12 - 13 | 2478 | 78 | |
| GenBank mRNA | AF045160.1 | - | 12 - 13 | 2242 | 78 | |
| AK141185.1 | - | 12 - 13 | 2213 | 78 | ||
| AK158430.1 | - | 12 - 13 | 2642 | 78 | ||
| BC057870.1 | - | 12 - 13 | 2200 | 5032291 | 78 | |
| D89787.1 | - | 12 - 13 | 2440 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epas1
Genomic Map