Assay Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010137.3 | NP_034267.3 | 14 - 15 | 2723 | 64 | |
| GenBank mRNA | AK136911.1 | - | 14 - 15 | 2727 | 64 | |
| AK158430.1 | - | 14 - 15 | 2887 | 64 | ||
| BC057870.1 | - | 14 - 15 | 2445 | 5032291 | 64 | |
| D89787.1 | - | 14 - 15 | 2685 | 64 | ||
| U81983.1 | - | 14 - 15 | 2406 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epas1
Genomic Map