Assay Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010137.3 | NP_034267.3 | 1 - 2 | 451 | 69 | |
| GenBank mRNA | AF045160.1 | - | 1 - 2 | 215 | 69 | |
| AK136911.1 | - | 1 - 2 | 451 | 69 | ||
| AK141185.1 | - | 1 - 2 | 188 | 69 | ||
| AK158430.1 | - | 1 - 2 | 615 | 69 | ||
| BC057870.1 | - | 1 - 2 | 173 | 5032291 | 69 | |
| D89787.1 | - | 1 - 2 | 413 | 69 | ||
| U81983.1 | - | 1 - 2 | 131 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF045160.1, AK158430.1, BC057870.1, D89787.1, NM_010137.3, U81983.1 transcripts.
Set Membership: |
|

Epas1
Genomic Map