Assay Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010137.3 | NP_034267.3 | 8 - 9 | 1451 | 68 | |
| GenBank mRNA | AF045160.1 | - | 8 - 9 | 1215 | 68 | |
| AK136911.1 | - | 8 - 9 | 1455 | 68 | ||
| AK141185.1 | - | 8 - 9 | 1186 | 68 | ||
| AK158430.1 | - | 8 - 9 | 1615 | 68 | ||
| BC057870.1 | - | 8 - 9 | 1173 | 5032291 | 68 | |
| D89787.1 | - | 8 - 9 | 1413 | 68 | ||
| U81983.1 | - | 8 - 9 | 1134 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epas1
Genomic Map