Assay Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163505.1 | NP_001156977.1 | 9 - 10 | 1242 | 93 | |
| NM_146091.4 | NP_666203.3 | 9 - 10 | 1144 | 93 | ||
| GenBank mRNA | AK028842.1 | - | 9 - 10 | 1005 | 93 | |
| AK030660.1 | - | 9 - 10 | 1004 | 93 | ||
| AK077752.1 | - | 9 - 10 | 1007 | 93 | ||
| AK147720.1 | - | 9 - 10 | 1102 | 93 | ||
| AK169476.1 | - | 9 - 10 | 1004 | 93 | ||
| AK170550.1 | - | 10 - 11 | 1095 | 93 | ||
| BC002149.1 | - | 6 - 7 | 534 | 93 | ||
| BC017138.1 | - | 9 - 10 | 993 | 4485115 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atl3
Genomic Map