Assay Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163505.1 | NP_001156977.1 | 2 - 3 | 537 | 102 | |
| NM_146091.4 | NP_666203.3 | 2 - 3 | 439 | 102 | ||
| GenBank mRNA | AK028842.1 | - | 2 - 3 | 300 | 102 | |
| AK030660.1 | - | 2 - 3 | 299 | 102 | ||
| AK077752.1 | - | 2 - 3 | 302 | 102 | ||
| AK147720.1 | - | 2 - 3 | 397 | 102 | ||
| AK169476.1 | - | 2 - 3 | 299 | 102 | ||
| AK170550.1 | - | 2 - 3 | 343 | 102 | ||
| BC017138.1 | - | 2 - 3 | 288 | 4485115 | 102 | |
| CA318221.1 | - | 2 - 3 | 439 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atl3
Genomic Map