Assay Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163505.1 | NP_001156977.1 | 3 - 4 | 683 | 95 | |
| NM_146091.4 | NP_666203.3 | 3 - 4 | 585 | 95 | ||
| GenBank mRNA | AK028842.1 | - | 3 - 4 | 446 | 95 | |
| AK030660.1 | - | 3 - 4 | 445 | 95 | ||
| AK077752.1 | - | 3 - 4 | 448 | 95 | ||
| AK147720.1 | - | 3 - 4 | 543 | 95 | ||
| AK169476.1 | - | 3 - 4 | 445 | 95 | ||
| AK170550.1 | - | 3 - 4 | 489 | 95 | ||
| BC017138.1 | - | 3 - 4 | 434 | 4485115 | 95 | |
| CA318221.1 | - | 3 - 4 | 585 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atl3
Genomic Map