Assay Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163505.1 | NP_001156977.1 | 5 - 6 | 834 | 133 | |
| NM_146091.4 | NP_666203.3 | 5 - 6 | 736 | 133 | ||
| GenBank mRNA | AK028842.1 | - | 5 - 6 | 597 | 133 | |
| AK030660.1 | - | 5 - 6 | 596 | 133 | ||
| AK077752.1 | - | 5 - 6 | 599 | 133 | ||
| AK147720.1 | - | 5 - 6 | 694 | 133 | ||
| AK169476.1 | - | 5 - 6 | 596 | 133 | ||
| AK170550.1 | - | 5 - 6 | 640 | 133 | ||
| BC002149.1 | - | 2 - 3 | 126 | 133 | ||
| BC017138.1 | - | 5 - 6 | 585 | 4485115 | 133 | |
More Information
Set Membership: |
|

Atl3
Genomic Map