Assay Details
| Gene Symbol: Atl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163505.1 | NP_001156977.1 | 6 - 7 | 893 | 107 | |
| NM_146091.4 | NP_666203.3 | 6 - 7 | 795 | 107 | ||
| GenBank mRNA | AK028842.1 | - | 6 - 7 | 656 | 107 | |
| AK030660.1 | - | 6 - 7 | 655 | 107 | ||
| AK077752.1 | - | 6 - 7 | 658 | 107 | ||
| AK147720.1 | - | 6 - 7 | 753 | 107 | ||
| AK169476.1 | - | 6 - 7 | 655 | 107 | ||
| AK170550.1 | - | 6 - 7 | 699 | 107 | ||
| BC002149.1 | - | 3 - 4 | 185 | 107 | ||
| BC017138.1 | - | 6 - 7 | 644 | 4485115 | 107 | |
More Information
Set Membership: |
|

Atl3
Genomic Map