Assay Details
| Gene Symbol: Slc22a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144785.2 | NP_659034.1 | 3 - 4 | 773 | 96 | |
| GenBank mRNA | AK052752.1 | - | 3 - 4 | 743 | 96 | |
| BC021384.1 | - | 3 - 4 | 739 | 5064542 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a19
Genomic Map