Assay Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016737.2 | NP_058017.1 | 6 - 7 | 940 | 73 | |
| GenBank mRNA | AK088494.1 | - | 6 - 7 | 882 | 73 | |
| AK149493.1 | - | 6 - 7 | 856 | 73 | ||
| AK161645.1 | - | 6 - 7 | 859 | 73 | ||
| AK167273.1 | - | 6 - 7 | 874 | 73 | ||
| AK168181.1 | - | 6 - 7 | 856 | 73 | ||
| AK189522.1 | - | 2 - 3 | 285 | 73 | ||
| BC003794.1 | - | 6 - 7 | 849 | 3590457 | 73 | |
| U27830.1 | - | 6 - 7 | 857 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stip1
Genomic Map