Assay Details
| Gene Symbol: Stip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016737.2 | NP_058017.1 | 7 - 8 | 1036 | 131 | |
| GenBank mRNA | AK075988.1 | - | 7 - 8 | 974 | 131 | |
| AK088494.1 | - | 7 - 8 | 978 | 131 | ||
| AK149493.1 | - | 7 - 8 | 952 | 131 | ||
| AK161645.1 | - | 7 - 8 | 955 | 131 | ||
| AK167273.1 | - | 7 - 8 | 970 | 131 | ||
| U27830.1 | - | 7 - 8 | 953 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stip1
Genomic Map