Assay Details
| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130449.2 | NP_569716.2 | 2 - 3 | 167 | 124 | |
| GenBank mRNA | AB078434.1 | - | 2 - 3 | 153 | 124 | |
| AK042772.1 | - | 2 - 3 | 168 | 124 | ||
| AK141928.1 | - | 2 - 3 | 153 | 124 | ||
| AK158366.1 | - | 2 - 3 | 189 | 124 | ||
| BC009162.1 | - | 2 - 3 | 156 | 124 | ||
| BC057936.1 | - | 2 - 3 | 156 | 5326386 | 124 | |
| BC060657.1 | - | 2 - 3 | 124 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Colec12
Genomic Map