Assay Details
| Gene Symbol: Colec12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130449.2 | NP_569716.2 | 5 - 6 | 1435 | 77 | |
| GenBank mRNA | AB038519.1 | - | 5 - 6 | 1406 | 77 | |
| AB078434.1 | - | 5 - 6 | 1421 | 77 | ||
| AK042772.1 | - | 5 - 6 | 1436 | 77 | ||
| AK141928.1 | - | 5 - 6 | 1421 | 77 | ||
| BC009162.1 | - | 5 - 6 | 1425 | 77 | ||
| BC048915.1 | - | 1 - 2 | 121 | 77 | ||
| BC057936.1 | - | 5 - 6 | 1424 | 5326386 | 77 | |
| BC060657.1 | - | 5 - 6 | 1413 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Colec12
Genomic Map