Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800007158.mm.1 |
NM_001038589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007158.mm.1 |
NM_001038589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007158.mm.1 |
NM_021522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007158.mm.1 |
NM_021522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007158.mm.1 |
TC1800000968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007157.mm.1 |
NM_001038589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007157.mm.1 |
NM_001038589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007157.mm.1 |
NM_021522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007157.mm.1 |
NM_021522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007157.mm.1 |
TC1800000968.mm.1 |
MTA TC |