Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021522.4 | NP_067497.2 | 3 - 4 | 420 | 80 | |
| GenBank mRNA | AB034633.1 | - | 3 - 4 | 199 | 80 | |
| AK011322.1 | - | 3 - 4 | 356 | 80 | ||
| AK029977.1 | - | 3 - 4 | 349 | 80 | ||
| AK045909.1 | - | 3 - 4 | 340 | 80 | ||
| AK160215.1 | - | 3 - 4 | 354 | 80 | ||
| AK170933.1 | - | 414 | 80 | |||
| AK172477.1 | - | 3 - 4 | 407 | 80 | ||
| BC005571.1 | - | 3 - 4 | 339 | 3157723 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170933.1 transcripts.
Set Membership: |
|

Usp14
Genomic Map