Assay Details
| Gene Symbol: Kdm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172132.2 | NP_742144.1 | 15 - 16 | 2474 | 76 | |
| GenBank mRNA | AK028962.1 | - | 4 - 5 | 642 | 76 | |
| AK043678.1 | - | 1 - 2 | 135 | 76 | ||
| AK129233.1 | - | 15 - 16 | 2412 | 76 | ||
| AK141879.1 | - | 15 - 16 | 2451 | 76 | ||
| BC005480.1 | - | 4 - 5 | 546 | 76 | ||
| BC007145.1 | - | 15 - 16 | 2444 | 3490671 | 76 | |
More Information
Set Membership: |
|

Kdm4b
Genomic Map