Assay Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023324.2 | NP_075813.2 | 4 - 5 | 729 | 105 | |
| GenBank mRNA | AF302503.1 | - | 3 - 4 | 951 | 105 | |
| AK151660.1 | - | 4 - 5 | 730 | 105 | ||
| AK152900.1 | - | 4 - 5 | 730 | 105 | ||
| AK152997.1 | - | 4 - 5 | 729 | 105 | ||
| AK153372.1 | - | 4 - 5 | 730 | 105 | ||
| AK167525.1 | - | 4 - 5 | 730 | 105 | ||
| BC016515.1 | - | 3 - 4 | 328 | 3600130 | 105 | |
More Information
Set Membership: |
|

Peli1
Genomic Map