Assay Details
| Gene Symbol: Macc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163136.1 | NP_001156608.1 | 2 - 3 | 2156 | 137 | |
| GenBank mRNA | AK028965.1 | - | 1 - 2 | 478 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42151 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426579 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111001 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 33719 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122626 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168003 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168003 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07769140 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122626 | - | - |
| Whole Mouse Genome G4122A | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243514 | AK028965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P243514 | NM_001163136.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09941132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10374810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10361616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09997105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09762176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10030631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10030625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09980799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10326528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10431369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10314349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09980797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28785164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1457044_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10399202 | - | - |
| Mouse Genome 430 2.0 Array | 1457044_at | - | - |
| Mouse Genome 430 2.0 Array | 1457044_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238455_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238455_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200009580.mm.1 | NM_001163136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009580.mm.1 | NM_001163136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009580.mm.1 | TC1200001335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200009582.mm.1 | NM_001163136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009582.mm.1 | NM_001163136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009582.mm.1 | TC1200001335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200009579.mm.1 | NM_001163136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009579.mm.1 | NM_001163136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009579.mm.1 | TC1200001335.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200009581.mm.1 | NM_001163136.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009581.mm.1 | NM_001163136.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200009581.mm.1 | TC1200001335.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457044_at | - | - |
| Mouse Expression Set 430 | 1457044_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457044_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457044_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 478376 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685733 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685733 | - | - |

Macc1
Genomic Map