Assay Details
| Gene Symbol: Ccng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009831.2 | NP_033961.1 | 4 - 5 | 887 | 74 | |
| GenBank mRNA | AK128957.1 | - | 4 - 5 | 875 | 74 | |
| AK136645.1 | - | 4 - 5 | 624 | 74 | ||
| AK143114.1 | - | 4 - 5 | 878 | 74 | ||
| AK147026.1 | - | 4 - 5 | 875 | 74 | ||
| AK153392.1 | - | 4 - 5 | 819 | 74 | ||
| AK159429.1 | - | 4 - 5 | 817 | 74 | ||
| AK159831.1 | - | 4 - 5 | 822 | 74 | ||
| AK160411.1 | - | 4 - 5 | 862 | 74 | ||
| AK167052.1 | - | 4 - 5 | 819 | 74 | ||
| AK167486.1 | - | 4 - 5 | 814 | 74 | ||
| AK170397.1 | - | 4 - 5 | 879 | 74 | ||
| AK178990.1 | - | 3 - 4 | 400 | 74 | ||
| AK186078.1 | - | 2 - 3 | 351 | 74 | ||
| AK219604.1 | - | 2 - 3 | 352 | 74 | ||
| BC005534.1 | - | 3 - 4 | 537 | 74 | ||
| BC006609.1 | - | 4 - 5 | 770 | 74 | ||
| L49507.1 | - | 4 - 5 | 830 | 74 | ||
| Z37110.1 | - | 4 - 5 | 830 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccng1
Genomic Map