Assay Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013552.2 | NP_038580.2 | 13 - 14 | 1859 | 66 | |
| GenBank mRNA | AF031932.1 | - | 13 - 14 | 1844 | 66 | |
| AF079222.1 | - | 14 - 15 | 1808 | 66 | ||
| AK143123.1 | - | 13 - 14 | 1859 | 66 | ||
| AK163410.1 | - | 13 - 14 | 1739 | 66 | ||
| BC021427.1 | - | 1983 | 5035341 | 66 | ||
| X64550.1 | - | 9 - 10 | 1472 | 66 | ||
More Information
Set Membership: |
|

Hmmr
Genomic Map