Assay Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013552.2 | NP_038580.2 | 3 - 4 | 307 | 110 | |
| GenBank mRNA | AF031932.1 | - | 3 - 4 | 292 | 110 | |
| AK020144.1 | - | 3 - 4 | 262 | 110 | ||
| AK041617.1 | - | 3 - 4 | 287 | 110 | ||
| AK143123.1 | - | 3 - 4 | 307 | 110 | ||
| AK163410.1 | - | 3 - 4 | 233 | 110 | ||
| AK164410.1 | - | 3 - 4 | 281 | 110 | ||
| CN455388.1 | - | 3 - 4 | 311 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmmr
Genomic Map