Assay Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013552.2 | NP_038580.2 | 4 - 5 | 352 | 90 | |
| GenBank mRNA | AF031932.1 | - | 4 - 5 | 337 | 90 | |
| AK020144.1 | - | 4 - 5 | 307 | 90 | ||
| AK041617.1 | - | 4 - 5 | 332 | 90 | ||
| AK143123.1 | - | 4 - 5 | 352 | 90 | ||
| AK163410.1 | - | 4 - 5 | 278 | 90 | ||
| AK164410.1 | - | 4 - 5 | 326 | 90 | ||
| CN455388.1 | - | 4 - 5 | 356 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hmmr
Genomic Map