Assay Details
| Gene Symbol: Ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009212.2 | NP_033238.2 | 4 - 5 | 772 | 61 | |
| GenBank mRNA | AK142456.1 | - | 4 - 5 | 576 | 61 | |
| AK153763.1 | - | 4 - 5 | 733 | 61 | ||
| AK154805.1 | - | 4 - 5 | 769 | 61 | ||
| AK155899.1 | - | 4 - 5 | 769 | 61 | ||
| BC066215.1 | - | 4 - 5 | 715 | 61 | ||
| BC083142.1 | - | 4 - 5 | 577 | 61 | ||
| BC133681.1 | - | 4 - 5 | 580 | 61 | ||
| L10075.1 | - | 4 - 5 | 668 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ighmbp2
Genomic Map