Assay Details
| Gene Symbol: Ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009212.2 | NP_033238.2 | 8 - 9 | 1457 | 107 | |
| GenBank mRNA | AK142456.1 | - | 8 - 9 | 1261 | 107 | |
| AK153763.1 | - | 8 - 9 | 1418 | 107 | ||
| AK154805.1 | - | 8 - 9 | 1454 | 107 | ||
| AK155899.1 | - | 8 - 9 | 1454 | 107 | ||
| BC052025.1 | - | 7 - 8 | 1329 | 107 | ||
| BC060085.1 | - | 7 - 8 | 1329 | 107 | ||
| BC066215.1 | - | 8 - 9 | 107 | |||
| BC083142.1 | - | 8 - 9 | 1262 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ighmbp2
Genomic Map