Assay Details
| Gene Symbol: Rps6kb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021485.2 | NP_067460.1 | 2 - 3 | 120 | 77 | |
| GenBank mRNA | AK014412.1 | - | 2 - 3 | 199 | 77 | |
| AK090093.1 | - | 175 | 77 | |||
| AK156361.1 | - | 2 - 3 | 173 | 77 | ||
| BC051631.1 | - | 2 - 3 | 175 | 77 | ||
| BC066842.1 | - | 2 - 3 | 191 | 30468251 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps6kb2
Genomic Map