Assay Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019449.2 | NP_062322.2 | 9 - 10 | 1433 | 70 | |
| GenBank mRNA | AK030488.1 | - | 9 - 10 | 1393 | 70 | |
| AK155556.1 | - | 9 - 10 | 1391 | 70 | ||
| AK165060.1 | - | 9 - 10 | 1393 | 70 | ||
| BC018388.1 | - | 9 - 10 | 1400 | 4209296 | 70 | |
| BC023731.1 | - | 3 - 4 | 858 | 5352340 | 70 | |
| BC025587.1 | - | 2 - 3 | 335 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc93b1
Genomic Map