Assay Details
| Gene Symbol: Ptprcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016933.3 | NP_058629.2 | 1 - 2 | 64 | 68 | |
| GenBank mRNA | AI323196.1 | - | 1 - 2 | 36 | 68 | |
| AK088334.1 | - | 1 - 2 | 62 | 68 | ||
| AK165203.1 | - | 1 - 2 | 62 | 68 | ||
| BB840651.1 | - | 1 - 2 | 62 | 68 | ||
| BC013273.1 | - | 1 - 2 | 63 | 4208992 | 68 | |
| BC100560.1 | - | 1 - 2 | 45 | 832557 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088334.1, AK165203.1, BC013273.1, BC100560.1, NM_016933.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493190 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132036 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146989 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127526 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | U03856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | U03856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921430 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127526 | - | - |
| Whole Mouse Genome G4122A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | U03856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138799 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127526 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146989 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14371126 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14222828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14222824 | - | - |
| Agilent Mouse cDNA Microarray (G4104A) | 642720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14463643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14222822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14371126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31921424 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460374 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448511_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460371 | - | - |
| Mouse Genome 430 2.0 Array | 1448511_at | - | - |
| Mouse Genome 430 2.0 Array | 1448511_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19265_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19265_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa13780_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000461.mm.1 | NM_016933.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000461.mm.1 | NM_016933.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000461.mm.1 | TC1900000031.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000462.mm.1 | NM_016933.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000462.mm.1 | NM_016933.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000462.mm.1 | TC1900000031.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448511_at | - | - |
| Mouse Expression Set 430 | 1448511_PM_at | - | - |
| Murine Genome U74v2 Set | 99478_PM_at | - | - |
| Murine Genome U74v2 Set | 99478_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448511_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448511_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448511_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448511_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6650075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652719 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2652719 | - | - |
| mouse-6 v1.1 (Illumina) | scl52802.3.52_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52802.3.52_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650075 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2652719 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652719 | - | - |

Ptprcap
Genomic Map