Assay Details
| Gene Symbol: Aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164828.1 | NP_001158300.1 | 3 - 4 | 476 | 119 | |
| NM_025651.4 | NP_079927.2 | 4 - 5 | 1993 | 119 | ||
| GenBank mRNA | AK003146.1 | - | 3 - 4 | 1330 | 119 | |
| AK006153.1 | - | 3 - 4 | 478 | 119 | ||
| AK030661.1 | - | 4 - 5 | 1864 | 119 | ||
| BC048866.1 | - | 3 - 4 | 454 | 119 | ||
| BC058377.1 | - | 4 - 5 | 1993 | 119 | ||
| BC098471.1 | - | 5 - 6 | 1878 | 30043996 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26797106 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281014 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283525 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158425 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26797135 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158425 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06912154 | - | - |
| Whole Mouse Genome G4122A | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P332003 | AK003146.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133983 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158425 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07498636 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07498635 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07667735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26797106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26797149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07498635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07488948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07784875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08124339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07488960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07488943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07969589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08124348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26797141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588423 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431270_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588419 | - | - |
| Mouse Genome 430 2.0 Array | 1431270_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1445012_at | - | - |
| Mouse Genome 430 2.0 Array | 1445012_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431270_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66595_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66595_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900010350.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010350.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010350.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010350.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010350.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010347.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010347.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010347.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010347.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010347.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010349.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010349.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010349.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010349.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010349.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010345.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010345.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010345.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010348.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010348.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010348.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010348.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010348.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010352.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010352.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010352.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010352.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010352.mm.1 | TC0900001325.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900010353.mm.1 | NM_001164828.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010353.mm.1 | NM_001164828.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010353.mm.1 | NM_025651.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010353.mm.1 | NM_025651.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900010353.mm.1 | TC0900001325.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431270_PM_a_at | - | - |
| Mouse Expression Set 430 | 1431270_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431270_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431270_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431270_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431270_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 684845 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1090079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214472 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214472 | - | - |
| mouse-6 v1.1 (Illumina) | scl36523.6.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36523.6.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630756 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630756 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36523.6.1_28-S | - | - |

Aste1
Genomic Map