Assay Details
| Gene Symbol: Pik3r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081309.1 | NP_001074778.1 | 12 - 13 | 3370 | 73 | |
| GenBank mRNA | AK050333.1 | - | 12 - 13 | 3340 | 73 | |
| AK054559.1 | - | 12 - 13 | 3387 | 73 | ||
| AK087938.1 | - | 12 - 13 | 3361 | 73 | ||
| BC024083.1 | - | 9 - 10 | 1844 | 73 | ||
| BC157947.1 | - | 12 - 13 | 3195 | 9007281 | 73 | |
| BC158047.1 | - | 12 - 13 | 3195 | 9088160 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r4
Genomic Map