Assay Details
| Gene Symbol: Lonp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028782.2 | NP_083058.2 | 15 - 16 | 2318 | 69 | |
| GenBank mRNA | AK004820.1 | - | 15 - 16 | 2302 | 69 | |
| AK131910.1 | - | 15 - 16 | 2304 | 69 | ||
| AK157474.1 | - | 15 - 16 | 2297 | 69 | ||
| AK159302.1 | - | 15 - 16 | 2303 | 69 | ||
| AK160071.1 | - | 15 - 16 | 2303 | 69 | ||
| AK160076.1 | - | 15 - 16 | 2304 | 69 | ||
| AK161983.1 | - | 15 - 16 | 2303 | 69 | ||
| AY190302.1 | - | 15 - 16 | 2288 | 69 | ||
| BC057365.1 | - | 13 - 14 | 3712 | 69 | ||
| BC058735.1 | - | 13 - 14 | 3712 | 69 | ||
| BC092212.1 | - | 6 - 7 | 685 | 69 | ||
More Information
Set Membership: |
|

Lonp1
Genomic Map