Assay Details
| Gene Symbol: Clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017393.2 | NP_059089.1 | 2 - 3 | 324 | 89 | |
| GenBank mRNA | AJ005253.1 | - | 2 - 3 | 292 | 89 | |
| AK002238.1 | - | 2 - 3 | 263 | 89 | ||
| AK004024.1 | - | 2 - 3 | 273 | 89 | ||
| AK145765.1 | - | 2 - 3 | 286 | 89 | ||
| AK147719.1 | - | 2 - 3 | 252 | 89 | ||
| AK168053.1 | - | 2 - 3 | 271 | 89 | ||
| BC001998.1 | - | 2 - 3 | 283 | 3487489 | 89 | |
| BY294002.1 | - | 2 - 3 | 325 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002238.1 transcripts.
Set Membership: |
|

Clpp
Genomic Map