Assay Details
| Gene Symbol: Clpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017393.2 | NP_059089.1 | 4 - 5 | 607 | 74 | |
| GenBank mRNA | AJ005253.1 | - | 4 - 5 | 575 | 74 | |
| AK002238.1 | - | 4 - 5 | 546 | 74 | ||
| AK004024.1 | - | 4 - 5 | 556 | 74 | ||
| AK145765.1 | - | 4 - 5 | 569 | 74 | ||
| AK147719.1 | - | 4 - 5 | 535 | 74 | ||
| AK168053.1 | - | 4 - 5 | 554 | 74 | ||
| BC001998.1 | - | 4 - 5 | 566 | 3487489 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clpp
Genomic Map