Assay Details
| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019418.3 | NP_062291.1 | 1 - 2 | 326 | 105 | |
| GenBank mRNA | AB029155.1 | - | 1 - 2 | 337 | 105 | |
| AF123385.1 | - | 1 - 2 | 214 | 105 | ||
| AF227533.1 | - | 1 - 2 | 224 | 105 | ||
| BC125477.1 | - | 1 - 2 | 303 | 40129992 | 105 | |
| BC125479.1 | - | 1 - 2 | 303 | 40129994 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf14
Genomic Map