Assay Details
| Gene Symbol: Pcyt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177546.2 | NP_808214.1 | 4 - 5 | 720 | 88 | |
| NM_211138.1 | NP_997593.1 | 4 - 5 | 731 | 88 | ||
| GenBank mRNA | AK032027.1 | - | 4 - 5 | 433 | 88 | |
| AK053577.1 | - | 4 - 5 | 705 | 88 | ||
| AK149304.1 | - | 4 - 5 | 721 | 88 | ||
| AY189949.1 | - | 4 - 5 | 568 | 88 | ||
| AY189950.1 | - | 4 - 5 | 720 | 88 | ||
| BC048917.1 | - | 4 - 5 | 602 | 6309383 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyt1b
Genomic Map