Assay Details
| Gene Symbol: Gdpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB048364.1 | - | 9 - 10 | 990 | 76 | |
| AK018634.1 | - | 9 - 10 | 817 | 76 | ||
| AK086508.1 | - | 9 - 10 | 983 | 76 | ||
| BC038274.1 | - | 9 - 10 | 1060 | 4457335 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gdpd2
Genomic Map