Assay Details
| Gene Symbol: Slc9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033289.2 | NP_001028461.2 | 10 - 11 | 2142 | 91 | |
| GenBank mRNA | AK077026.1 | - | 10 - 11 | 2234 | 91 | |
| AK138779.1 | - | 10 - 11 | 2520 | 91 | ||
| BC103685.1 | - | 10 - 11 | 2142 | 40048593 | 91 | |
| BC104737.1 | - | 10 - 11 | 2142 | 40048591 | 91 | |
| BC105676.1 | - | 10 - 11 | 2141 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a2
Genomic Map