Assay Details
| Gene Symbol: Slc9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033289.2 | NP_001028461.2 | 4 - 5 | 1393 | 67 | |
| GenBank mRNA | AK077026.1 | - | 4 - 5 | 1485 | 67 | |
| AK138779.1 | - | 4 - 5 | 1770 | 67 | ||
| BC103685.1 | - | 4 - 5 | 1393 | 40048593 | 67 | |
| BC104737.1 | - | 4 - 5 | 1393 | 40048591 | 67 | |
| BC105676.1 | - | 4 - 5 | 1392 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a2
Genomic Map