Assay Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177084.3 | NP_796058.1 | 2 - 3 | 1007 | 92 | |
| GenBank mRNA | AK085681.1 | - | 2 - 3 | 1033 | 92 | |
| AK141534.1 | - | 2 - 3 | 1008 | 92 | ||
| BC120543.1 | - | 2 - 3 | 40129466 | 92 | ||
More Information
Set Membership: |
|

Slc9a4
Genomic Map