Assay Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177084.3 | NP_796058.1 | 8 - 9 | 2003 | 64 | |
| GenBank mRNA | AK085681.1 | - | 8 - 9 | 2029 | 64 | |
| AK141534.1 | - | 8 - 9 | 2004 | 64 | ||
| AK170522.1 | - | 6 - 7 | 842 | 64 | ||
| BC120543.1 | - | 8 - 9 | 1921 | 40129466 | 64 | |
More Information
Set Membership: |
|

Slc9a4
Genomic Map