Assay Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177084.3 | NP_796058.1 | 9 - 10 | 2099 | 99 | |
| GenBank mRNA | AK085681.1 | - | 9 - 10 | 2125 | 99 | |
| AK141534.1 | - | 9 - 10 | 2100 | 99 | ||
| AK170522.1 | - | 7 - 8 | 938 | 99 | ||
| BC120543.1 | - | 9 - 10 | 2017 | 40129466 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a4
Genomic Map