Assay Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172499.2 | NP_766087.1 | 1 - 2 | 171 | 57 | |
| GenBank mRNA | AK029874.1 | - | 1 - 2 | 172 | 57 | |
| AK170820.1 | - | 1 - 2 | 172 | 57 | ||
| BC057633.1 | - | 1 - 2 | 177 | 4035998 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd9
Genomic Map