Assay Details
| Gene Symbol: Tbc1d15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025706.3 | NP_079982.3 | 15 - 16 | 1795 | 119 | |
| GenBank mRNA | AK014477.1 | - | 15 - 16 | 1727 | 119 | |
| AK014795.1 | - | 10 - 11 | 1178 | 119 | ||
| AK078875.1 | - | 15 - 16 | 1741 | 119 | ||
| AK135905.1 | - | 11 - 12 | 1271 | 119 | ||
| AK147085.1 | - | 15 - 16 | 1758 | 119 | ||
| AK162115.1 | - | 4 - 5 | 393 | 119 | ||
| BC032087.1 | - | 15 - 16 | 1723 | 119 | ||
| BC053395.1 | - | 15 - 16 | 1776 | 30054233 | 119 | |
| DQ054831.1 | - | 15 - 16 | 1726 | 119 | ||
More Information
Set Membership: |
|

Tbc1d15
Genomic Map