Assay Details
| Gene Symbol: Tbc1d15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025706.3 | NP_079982.3 | 9 - 10 | 1176 | 103 | |
| GenBank mRNA | AK014477.1 | - | 9 - 10 | 1108 | 103 | |
| AK014795.1 | - | 4 - 5 | 560 | 103 | ||
| AK078875.1 | - | 9 - 10 | 1122 | 103 | ||
| AK135905.1 | - | 5 - 6 | 651 | 103 | ||
| AK147085.1 | - | 9 - 10 | 1139 | 103 | ||
| BC032087.1 | - | 9 - 10 | 1104 | 103 | ||
| BC053395.1 | - | 9 - 10 | 1157 | 30054233 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbc1d15
Genomic Map