Assay Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026565.2 | NP_080841.1 | 7 - 8 | 613 | 65 | |
| GenBank mRNA | AK012398.1 | - | 7 - 8 | 659 | 65 | |
| AK012804.1 | - | 7 - 8 | 654 | 65 | ||
| AK079141.1 | - | 7 - 8 | 665 | 65 | ||
| BC024334.1 | - | 7 - 8 | 613 | 5036743 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apool
Genomic Map