Assay Details
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027166.5 | NP_081442.1 | 1 - 2 | 89 | 55 | |
GenBank mRNA | AK010201.1 | - | 1 - 2 | 55 | 55 | |
AK046281.1 | - | 1 - 2 | 88 | 55 | ||
AK049096.1 | - | 1 - 2 | 90 | 55 | ||
AK049644.1 | - | 1 - 2 | 88 | 55 | ||
AK137446.1 | - | 1 - 2 | 61 | 55 | ||
AK165943.1 | - | 1 - 2 | 91 | 55 | ||
AK167998.1 | - | 1 - 2 | 105 | 55 | ||
AK169241.1 | - | 1 - 2 | 92 | 55 | ||
AK169986.1 | - | 1 - 2 | 63 | 55 | ||
AK171010.1 | - | 1 - 2 | 90 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010201.1, AK046281.1, AK049096.1, AK049644.1, AK137446.1, AK165943.1, AK167998.1, AK169241.1, AK169986.1, AK171010.1, NM_027166.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
