Assay Details
| Gene Symbol: Rfx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145660.1 | NP_001139132.1 | 14 - 15 | 1643 | 75 | |
| GenBank mRNA | AK133372.1 | - | 14 - 15 | 1655 | 75 | |
| BG808411.1 | - | 1 - 2 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
endocrine pancreas development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
biological_process
endocrine pancreas development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096189 | AK133372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096189 | BG808411.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096189 | NM_001145660.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48413 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155545 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 468979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AB026489.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK002557.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK004710.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK039491.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK075800.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK075813.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK084801.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK169007.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | AK170057.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | BC065778.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | BC070458.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | NM_023055.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914738 | NM_023449.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932096 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455426 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 949879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | AK004154.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | AK141496.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | AK153059.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | AK160695.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | BC038324.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | BC091734.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380377 | NM_054099.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74259 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096189 | AK133372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096189 | BG808411.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096189 | NM_001145660.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00050529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00050481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04071715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04071856 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04071776 | - | - |
| Whole Mouse Genome G4122A | A_52_P832709 | AK133372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P832709 | NM_001145660.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04071856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00329627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00651733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00348484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00986312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00329628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00071892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167013 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00391667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00294869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00120500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00345781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00120504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00345784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00126087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00177962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00120513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00294871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00914951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00073702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00963549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00708416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00271918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00071898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00963547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00900297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00294864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20167091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10354217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354218 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435724_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354216 | - | - |
| Mouse Genome 430 2.0 Array | 1435724_at | - | - |
| Mouse Genome 430 2.0 Array | 1435724_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 619289_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 619289_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100018067.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018067.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018067.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018053.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018053.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018053.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018058.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018058.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018058.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018060.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018060.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018060.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018066.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018066.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018066.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018056.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018056.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018056.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018051.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018051.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018051.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018061.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018061.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018061.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018065.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018065.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018065.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018054.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018054.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018054.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018064.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018064.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018064.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018052.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018052.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018052.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018063.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018063.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018063.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018055.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018055.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018055.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018057.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018057.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018057.mm.1 | TC0100002242.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100018059.mm.1 | NM_001145660.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018059.mm.1 | NM_001145660.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100018059.mm.1 | TC0100002242.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435724_PM_at | - | - |
| Mouse Expression Set 430 | 1435724_at | - | - |
| Murine Genome U74v2 Set | 166383_PM_r_at | - | - |
| Murine Genome U74v2 Set | 166383_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435724_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435724_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 359513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246518 | - | - |

Rfx8
Genomic Map