Assay Details
| Gene Symbol: Jph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020604.2 | NP_065629.1 | 4 - 5 | 2189 | 74 | |
| GenBank mRNA | AB024445.1 | - | 4 - 5 | 2098 | 74 | |
| BC120839.1 | - | 4 - 5 | 2189 | 40129762 | 74 | |
| BC137669.1 | - | 4 - 5 | 2189 | 8860688 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jph1
Genomic Map