Assay Details
| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195732.1 | NP_001182661.1 | 1 - 2 | 183 | 115 | |
| GenBank mRNA | AK087278.1 | - | 1 - 2 | 59 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam150a
Genomic Map